N-α-Formyl-D-leucine ≥90%

Supplier: Matrix Scientific

Synonyms: N-Formyl-D-leucine

054023-500MG
101905-880EA 203.08 USD
101905-880
N-α-Formyl-D-leucine ≥90%
N-alpha-Formyl-D-leucine

Hazard: Irritant

Formula: C₇H₁₃NO₃
MW: 159.185 g/mol
MDL Number: MFCD08275788
CAS Number: 44978-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR