N,N-Diallyl-5-bromo-2-pyridinamine

Supplier: Matrix Scientific

052140-500MG 052140-2.5G
101898-450EA 364.62 USD
101898-450 101898-452
N,N-Diallyl-5-bromo-2-pyridinamine
N,N-Diallyl-5-bromo-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₃BrN₂
MW: 253.15 g/mol
MDL Number: MFCD13561939
CAS Number: 309977-78-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR