Methyl (2S,4S)-4-(2-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051034-2.5G
101894-030EA 838.46 USD
101894-030
Methyl (2S,4S)-4-(2-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-propylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13561404
CAS Number: 1354487-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR