4-[(4-Isopropylbenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050629-2.5G
101892-418EA 838.46 USD
101892-418
4-[(4-Isopropylbenzyl)oxy]piperidine hydrochloride
4-[(4-Isopropylbenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13561025
CAS Number: 1220033-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR