4-[(Pentyloxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050602-2.5G
101892-310EA 838.46 USD
101892-310
4-[(Pentyloxy)methyl]piperidine hydrochloride
4-[(Pentyloxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄ClNO
MW: 221.77 g/mol
MDL Number: MFCD13560999
CAS Number: 1220027-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR