4-Piperidinyl cyclobutanecarboxylate hydrochloride

Supplier: Matrix Scientific

049318-500MG 049318-2.5G
101887-176EA 364.62 USD
101887-176 101887-178
4-Piperidinyl cyclobutanecarboxylate hydrochloride
4-Piperidinyl cyclobutanecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO₂
MW: 219.71 g/mol
MDL Number: MFCD13559816
CAS Number: 1219948-61-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR