(3-Allyl-4-hydroxy-5-methoxybenzyl)formamide

Supplier: Matrix Scientific

048756-500MG
101885-090EA 269.23 USD
101885-090
(3-Allyl-4-hydroxy-5-methoxybenzyl)formamide
(3-Allyl-4-hydroxy-5-methoxybenzyl)formamide

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
MDL Number: MFCD12195849
CAS Number: 1201633-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR