4-sec-Butylaniline

Supplier: Matrix Scientific

Synonyms: 4-(sec-Butyl)aniline

048749-500MG
101885-076EA 24.62 USD
101885-076
4-sec-Butylaniline
4-sec-Butylaniline

Hazard: Toxic

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 238 °C
Density: 0.94
Flash Pt: 107 °C
MDL Number: MFCD00010223
CAS Number: 30273-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR