Ethyl-2-cyano-5-(trifluoromethyl)nicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-cyano-5-(trifluoromethyl)nicotinate

048692-500MG 048692-1G
101884-892EA 921.54 USD
101884-892 101884-894
Ethyl-2-cyano-5-(trifluoromethyl)nicotinate ≥95%
Ethyl-2-cyano-5-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O₂
MW: 244.17 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD13193103
CAS Number: 1192263-82-3

Order Now

Specification Test Results

Melting point 55-57°C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR