Methyl-5-chloro-2-methoxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-chloro-2-methoxynicotinate, Methyl 5-chloro-2-methoxy-3-pyridinecarboxylate

046650-1G
101876-412EA 246.15 USD
101876-412
Methyl-5-chloro-2-methoxynicotinate ≥95%
Methyl-5-chloro-2-methoxynicotinate

Hazard: Irritant

Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
MDL Number: MFCD12025883
CAS Number: 82060-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR