Methyl-4-(5-nitro-2-pyridinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044355-500MG 044355-1G 044355-5G
101865-436EA 556.92 USD
101865-436 101865-438 101865-440
Methyl-4-(5-nitro-2-pyridinyl)benzenecarboxylate ≥95%
Methyl-4-(5-nitro-2-pyridinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₄
MW: 258.24 g/mol
Melting Pt: 198…200 °C
MDL Number: MFCD06200961
CAS Number: 223127-51-3

Order Now

Specification Test Results

Melting point 198-200°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR