Ethyl-2-[(2-fluorobenzoyl)amino]acetate ≥95%

Supplier: Matrix Scientific

043497-500MG 043497-1G 043497-5G
101861-110EA 181.54 USD
101861-110 101861-112 101861-114
Ethyl-2-[(2-fluorobenzoyl)amino]acetate ≥95%
Ethyl-2-[(2-fluorobenzoyl)amino]acetate

Hazard: Irritant

Formula: C₁₁H₁₂FNO₃
MW: 225.22 g/mol
Melting Pt: 40…42 °C
MDL Number: MFCD00793744
CAS Number: 304657-05-4

Order Now

Specification Test Results

Melting point 40-42°C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR