3-Fluoro-4-(N-morpholino)phenol

Supplier: Matrix Scientific

041148-250MG
101850-900EA 461.54 USD
101850-900
3-Fluoro-4-(N-morpholino)phenol
3-Fluoro-4-(N-morpholino)phenol

Hazard: Irritant

Formula: C₁₀H₁₂FNO₂
MW: 197.21 g/mol
MDL Number: MFCD11974203
CAS Number: 1171917-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR