2-Piperidin-3-yl-1H-pyrrolo[2,3-b]pyridine dihydrochloride

Supplier: Matrix Scientific

041072-250MG
101850-744EA 653.85 USD
101850-744
2-Piperidin-3-yl-1H-pyrrolo[2,3-b]pyridine dihydrochloride
2-Piperidin-3-yl-1H-pyrrolo[2,3-b]pyridine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂N₃
MW: 274.2 g/mol
MDL Number: MFCD12028518
CAS Number: 1185303-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR