3-(5-{[(2-Fluorophenyl)amino]carbonyl}-1,3,4-thiadiazol-2-yl)propanoic acid

Supplier: Matrix Scientific

040532-500MG
101849-698EA 387.69 USD
101849-698
3-(5-{[(2-Fluorophenyl)amino]carbonyl}-1,3,4-thiadiazol-2-yl)propanoic acid
3-(5-{[(2-Fluorophenyl)amino]carbonyl}-1,3,4-thiadiazol-2-yl)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₀FN₃O₃S
MW: 295.29 g/mol
MDL Number: MFCD12028242
CAS Number: 1142202-72-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR