[(2,3-Dimethylphenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

040470-500MG
101849-576EA 146.15 USD
101849-576
[(2,3-Dimethylphenyl)amino](oxo)acetic acid
[(2,3-Dimethylphenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD09864574
CAS Number: 959241-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR