4-(5-Amino-1,3,4-thiadiazol-2-yl)-1-(2-methylphenyl)pyrrolidin-2-one

Supplier: Matrix Scientific

040393-500MG
101849-422EA 255.38 USD
101849-422
4-(5-Amino-1,3,4-thiadiazol-2-yl)-1-(2-methylphenyl)pyrrolidin-2-one
4-(5-Amino-1,3,4-thiadiazol-2-yl)-1-(2-methylphenyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₃H₁₄N₄OS
MDL Number: MFCD12028221
CAS Number: 1142202-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR