3-Cyclohexyl-4-(hydroxyamino)-5,5-dimethyl-1,3-thiazolidine-2-thione

Supplier: Matrix Scientific

040367-500MG
101849-370EA 290.77 USD
101849-370
3-Cyclohexyl-4-(hydroxyamino)-5,5-dimethyl-1,3-thiazolidine-2-thione
3-Cyclohexyl-4-(hydroxyamino)-5,5-dimethyl-1,3-thiazolidine-2-thione

Hazard: Irritant

Formula: C₁₁H₂₀N₂OS₂
MDL Number: MFCD12028215
CAS Number: 400756-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR