[{2-[4-(4-Fluorophenyl)piperazin-1-yl]-2-oxoethyl}(phenyl)amino]acetic acid

Supplier: Matrix Scientific

039187-500MG
101846-992EA 193.85 USD
101846-992
[{2-[4-(4-Fluorophenyl)piperazin-1-yl]-2-oxoethyl}(phenyl)amino]acetic acid
[{2-[4-(4-Fluorophenyl)piperazin-1-yl]-2-oxoethyl}(phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₂₀H₂₂FN₃O₃
MDL Number: MFCD12027691
CAS Number: 1142211-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR