{(4-Methoxyphenyl)[2-(4-methyl-1,4-diazepan-1-yl)-2-oxoethyl]amino}acetic acid

Supplier: Matrix Scientific

039181-500MG
101846-980EA 193.85 USD
101846-980
{(4-Methoxyphenyl)[2-(4-methyl-1,4-diazepan-1-yl)-2-oxoethyl]amino}acetic acid
{(4-Methoxyphenyl)[2-(4-methyl-1,4-diazepan-1-yl)-2-oxoethyl]amino}acetic acid

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₄
MW: 335.41 g/mol
MDL Number: MFCD12027685
CAS Number: 1142205-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR