[{2-Oxo-2-[(2-pyridin-2-ylethyl)amino]ethyl}(phenyl)amino]acetic acid

Supplier: Matrix Scientific

039117-500MG
101846-856EA 193.85 USD
101846-856
[{2-Oxo-2-[(2-pyridin-2-ylethyl)amino]ethyl}(phenyl)amino]acetic acid
[{2-Oxo-2-[(2-pyridin-2-ylethyl)amino]ethyl}(phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₇H₁₉N₃O₃
MDL Number: MFCD12027621
CAS Number: 1142204-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR