N-(4-Chlorophenyl)pyrrolidin-3-amine

Supplier: Matrix Scientific

039022-500MG
101846-668EA 484.62 USD
101846-668
N-(4-Chlorophenyl)pyrrolidin-3-amine
N-(4-Chlorophenyl)pyrrolidin-3-amine

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂
MDL Number: MFCD06740351
CAS Number: 356558-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR