2-Aminonicotinaldehyde thiosemicarbazone

Supplier: Matrix Scientific

038949-500MG
101846-524EA 193.85 USD
101846-524
2-Aminonicotinaldehyde thiosemicarbazone
2-Aminonicotinaldehyde thiosemicarbazone

Hazard: Irritant

Formula: C₇H₉N₅S
MDL Number: MFCD12027515
CAS Number: 131418-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR