2-[(4-Aminophenyl)thio]-N-benzylacetamide

Supplier: Matrix Scientific

038909-500MG
101846-452EA 193.85 USD
101846-452
2-[(4-Aminophenyl)thio]-N-benzylacetamide
2-[(4-Aminophenyl)thio]-N-benzylacetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂OS
MDL Number: MFCD03551677
CAS Number: 710965-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR