3-(4-Methoxyphenyl)-2-methyl-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

Supplier: Matrix Scientific

038579-500MG
101845-788EA 193.85 USD
101845-788
3-(4-Methoxyphenyl)-2-methyl-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid
3-(4-Methoxyphenyl)-2-methyl-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₇NO₄
MDL Number: MFCD12027401
CAS Number: 442858-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR