Ethyl-3-isocyanatobenzoate

Supplier: Matrix Scientific

Synonyms: 3-(Ethoxycarbonyl)phenyl isocyanate

038315-500MG
101845-256EA 193.85 USD
101845-256
Ethyl-3-isocyanatobenzoate
Ethyl 3-isocyanatobenzoate

Hazard: Irritant

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
MDL Number: MFCD00013865
CAS Number: 67531-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR