Ethyl-3-pyrrolidin-1-ylpropanoate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(pyrrolidin-1-yl)propanoate

037261-500MG
101843-006EA 230.77 USD
101843-006
Ethyl-3-pyrrolidin-1-ylpropanoate
Ethyl-3-pyrrolidin-1-ylpropanoate

Hazard: Irritant

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
MDL Number: MFCD00020861
CAS Number: 6317-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR