5-(3-Aminothieno[2,3-b]pyridin-2-yl)-4-cyclopentyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

036769-500MG
101842-044EA 290.77 USD
101842-044
5-(3-Aminothieno[2,3-b]pyridin-2-yl)-4-cyclopentyl-4H-1,2,4-triazole-3-thiol
5-(3-Aminothieno[2,3-b]pyridin-2-yl)-4-cyclopentyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₄H₁₅N₅S₂
MDL Number: MFCD12026972
CAS Number: 1030433-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR