1-Ethyl-3,5-dimethyl-1H-pyrazole

Supplier: Matrix Scientific

Synonyms: 1-Ethyl-3,5-dimethyl-1H-pyrazole, 3,5-Dimethyl-1-ethyl-1H-pyrazole

036446-500MG
101841-382EA 387.69 USD
101841-382
1-Ethyl-3,5-dimethyl-1H-pyrazole
1-Ethyl-3,5-dimethyl-1H-pyrazole

Hazard: Irritant

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
MDL Number: MFCD02601834
CAS Number: 17629-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR