3-Ethyl-1,2,4-oxadiazole-5-carboxylic acid, potassium salt

Supplier: Matrix Scientific

036347-500MG
101841-180EA 387.69 USD
101841-180
3-Ethyl-1,2,4-oxadiazole-5-carboxylic acid, potassium salt
3-Ethyl-1,2,4-oxadiazole-5-carboxylic acid, potassium salt

Hazard: Irritant

Formula: C₅H₅N₂O₃K
MDL Number: MFCD22548409
CAS Number: 944906-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR