[(Cyclopropylmethyl)thio]acetic acid

Supplier: Matrix Scientific

036318-500MG
101841-122EA 193.85 USD
101841-122
[(Cyclopropylmethyl)thio]acetic acid
[(Cyclopropylmethyl)thio]acetic acid

Hazard: Irritant

Formula: C₆H₁₀O₂S
MDL Number: MFCD09864608
CAS Number: 959241-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR