3-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]-N-(cyclohexylmethyl)benzamide

Supplier: Matrix Scientific

036260-500MG
101841-004EA 484.62 USD
101841-004
3-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]-N-(cyclohexylmethyl)benzamide
3-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]-N-(cyclohexylmethyl)benzamide

Hazard: Irritant

Formula: C₁₇H₂₀ClN₃O₂
MDL Number: MFCD12026834
CAS Number: 1119450-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR