2-[(5-{[(3-Methylphenyl)amino]methyl}-4-phenyl-4H-1,2,4-triazol-3-yl)thio]acetohydrazide

Supplier: Matrix Scientific

036056-500MG
101840-598EA 387.69 USD
101840-598
2-[(5-{[(3-Methylphenyl)amino]methyl}-4-phenyl-4H-1,2,4-triazol-3-yl)thio]acetohydrazide
2-[(5-{[(3-Methylphenyl)amino]methyl}-4-phenyl-4H-1,2,4-triazol-3-yl)thio]acetohydrazide

Hazard: Irritant

Formula: C₁₈H₂₀N₆OS
MDL Number: MFCD02255657
CAS Number: 1071296-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR