Sklar Blue™ Sims Vaginal Retractor, OR Grade, Sklar
Supplier: Sklar
91-5200 91-5202 91-5212
10184-478EA 558.93 USD
10184-478 10184-480 10184-482
Sklar Blue™ Sims Vaginal Retractor, OR Grade, Sklar
Retractors
Sklar Blue™ Sims Vaginal Retractor is a handheld retractor used primarily for vaginal retraction and exposure in electrosurgical procedures
Sklar Blue™ electrosurgical instruments feature an insulated coating that prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. This retractor is available as a single or double-ended instrument with the blades always curved at right angles. This product has a single blade and a Sklar Grip handle that features ridges for comfortable finger placement and added control. There isMore Product Information