5-Bromo-2-formyl-3-methoxypyridine

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-methoxypicolinaldehyde

035193-250MG
101839-164EA 290.77 USD
101839-164
5-Bromo-2-formyl-3-methoxypyridine
5-Bromo-2-formyl-3-methoxypyridine

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
MDL Number: MFCD11857632
CAS Number: 1087659-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR