2-(2-Methoxy-1-naphthyl)-1-ethanamine hydrochloride

Supplier: Matrix Scientific

034893-500MG
101838-576EA 306.15 USD
101838-576
2-(2-Methoxy-1-naphthyl)-1-ethanamine hydrochloride
2-(2-Methoxy-1-naphthyl)-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆ClNO
MDL Number: MFCD11226484
CAS Number: 156482-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR