2-(Aminomethyl)-6-methoxyphenol hydrochloride

Supplier: Matrix Scientific

034886-500MG
101838-562EA 306.15 USD
101838-562
2-(Aminomethyl)-6-methoxyphenol hydrochloride
2-(Aminomethyl)-6-methoxyphenol hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
MDL Number: MFCD00465363
CAS Number: 141803-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR