Quinaldic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid

034842-100G
101838-480EA 113.85 USD
101838-480
Quinaldic acid ≥98%
Quinaldic acid

Hazard code(s): Irritant

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00006752
CAS Number: 93-10-7
EINECS: 202-218-3
Merck Index: 12,08227

Order Now

Specification Test Results

Melting point 156-159°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR