5-(1,1-Difluoroethyl)-1,2,3-trifluorobenzene ≥95%

Supplier: Matrix Scientific

034806-1G
101838-406EA 235.38 USD
101838-406
5-(1,1-Difluoroethyl)-1,2,3-trifluorobenzene ≥95%
5-(1,1-Difluoroethyl)-1,2,3-trifluorobenzene

Hazard: Irritant

Formula: C₈H₅F₅
MDL Number: MFCD11845820
CAS Number: 1138445-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR