1-Bromo-2-(3,3,3-trifluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

034708-1G
101838-196EA 273.85 USD
101838-196
1-Bromo-2-(3,3,3-trifluoropropyl)benzene ≥95%
1-Bromo-2-(3,3,3-trifluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₈BrF₃
MDL Number: MFCD11226673
CAS Number: 1099597-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR