1-Difluoromethyl-1H-pyrazole-4-sulfonyl chloride

Supplier: Matrix Scientific

034537-500MG
101837-850EA 330.77 USD
101837-850
1-Difluoromethyl-1H-pyrazole-4-sulfonyl chloride
1-Difluoromethyl-1H-pyrazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₄H₃ClF₂N₂O₂S
MDL Number: MFCD08558182
CAS Number: 1006320-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR