2-Phenylethanesulfonyl chloride

Supplier: Matrix Scientific

034526-1G
101837-828EA 150.77 USD
101837-828
2-Phenylethanesulfonyl chloride
2-Phenylethanesulfonyl chloride

Hazard: Irritant

Formula: C₈H₉ClO₂S
MW: 204.6739 g/mol
MDL Number: MFCD04117454
CAS Number: 4025-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR