1-(3-Bromopropyl)-2-methoxybenzene

Supplier: Matrix Scientific

034199-500MG
101837-178EA 453.85 USD
101837-178
1-(3-Bromopropyl)-2-methoxybenzene
1-(3-Bromopropyl)-2-methoxybenzene

Hazard: Irritant

Formula: C₁₀H₁₃BrO
MDL Number: MFCD00017896
CAS Number: 38011-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR