5-o-Tolyl-2H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

034173-500MG
101837-126EA 161.54 USD
101837-126
5-o-Tolyl-2H-pyrazole-3-carboxylic acid
5-o-Tolyl-2H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
MDL Number: MFCD08059586
CAS Number: 1140528-29-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR