3-Chloro-2-fluorobenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-fluoro-3-(trifluoromethyl)benzene

032384-1G
101833-612EA 120 USD
101833-612
3-Chloro-2-fluorobenzotrifluoride ≥95%
3-Chloro-2-fluorobenzotrifluoride

Hazard: Irritant

Formula: C₇H₃ClF₄
MW: 198.5500 g/mol
MDL Number: MFCD11100532
CAS Number: 1099597-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR