2-Bromo-5-iodo-3-nitropyridine

Supplier: Matrix Scientific

032320-500MG
101833-464EA 526.15 USD
101833-464
2-Bromo-5-iodo-3-nitropyridine
2-Bromo-5-iodo-3-nitropyridine

Hazard: Irritant

Formula: C₅H₂BrIN₂O₂
MW: 328.9 g/mol
MDL Number: MFCD10574979
CAS Number: 426463-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR