4-(Heptyloxy)-N-hexylaniline

Supplier: Matrix Scientific

031935-500MG
101832-694EA 438.46 USD
101832-694
4-(Heptyloxy)-N-hexylaniline
4-(Heptyloxy)-N-hexylaniline

Hazard: Irritant

Formula: C₁₉H₃₃NO
MDL Number: MFCD10687951
CAS Number: 1040689-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR