N-{2-[2-(sec-Butyl)phenoxy]butyl}-1-hexadecanamine

Supplier: Matrix Scientific

031552-500MG
101831-930EA 306.15 USD
101831-930
N-{2-[2-(sec-Butyl)phenoxy]butyl}-1-hexadecanamine
N-{2-[2-(sec-Butyl)phenoxy]butyl}-1-hexadecanamine

Hazard: Irritant

Formula: C₃₀H₅₅NO
MW: 445.78 g/mol
MDL Number: MFCD10687628
CAS Number: 1040688-22-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR