N-{2-[2-(tert-Butyl)phenoxy]ethyl}-2-isopropoxyaniline

Supplier: Matrix Scientific

031299-500MG
101831-424EA 306.15 USD
101831-424
N-{2-[2-(tert-Butyl)phenoxy]ethyl}-2-isopropoxyaniline
N-{2-[2-(tert-Butyl)phenoxy]ethyl}-2-isopropoxyaniline

Hazard: Irritant

Formula: C₂₁H₂₉NO₂
MDL Number: MFCD10687430
CAS Number: 1040683-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR