1-(1-Methyl-1H-pyrazol-5-yl)ethanone

Supplier: Matrix Scientific

030935-500MG
101831-134EA 173.85 USD
101831-134
1-(1-Methyl-1H-pyrazol-5-yl)ethanone
1-(1-Methyl-1H-pyrazol-5-yl)ethanone

Hazard: Irritant

Formula: C₆H₈N₂O
MDL Number: MFCD04968784
CAS Number: 137890-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR